Assay Details
| Gene Symbol: LOC4352780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765332.1 | XP_015620818.1 | 7 - 8 | 1293 | 71 | |
| GenBank mRNA | AK100618.1 | - | 7 - 8 | 1294 | 71 | |
| FJ940211.1 | - | 7 - 8 | 1047 | 71 | ||
More Information
Set Membership: |
|

LOC4352780
Genomic Map