Assay Details
| Gene Symbol: LOC4352796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765116.2 | XP_015620602.1 | 11 - 12 | 2082 | 92 | |
| GenBank mRNA | AK106725.1 | - | 3 - 4 | 603 | 92 | |
More Information
Set Membership: |
|

LOC4352796
Genomic Map