Assay Details
| Gene Symbol: LOC4352796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765116.2 | XP_015620602.1 | 13 - 14 | 2283 | 138 | |
| GenBank mRNA | AK106725.1 | - | 5 - 6 | 804 | 138 | |
More Information
Set Membership: |
|

LOC4352796
Genomic Map