Assay Details
| Gene Symbol: LOC4350053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761367.2 | XP_015616853.1 | 6 - 7 | 1076 | 75 | |
| GenBank mRNA | AK061267.1 | - | 7 - 8 | 760 | 75 | |
| AK069330.1 | - | 6 - 7 | 1082 | 75 | ||
| AK073964.1 | - | 824 | 75 | |||
| EF122490.1 | - | 6 - 7 | 846 | 75 | ||
| GQ848036.1 | - | 6 - 7 | 799 | 75 | ||
More Information
Set Membership: |
|

LOC4350053
Genomic Map