Assay Details
| Gene Symbol: LOC4350083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762471.2 | XP_015617957.1 | 6 - 7 | 741 | 70 | |
| XM_015762472.2 | XP_015617958.1 | 6 - 7 | 741 | 70 | ||
| GenBank mRNA | AK060326.1 | - | 6 - 7 | 730 | 70 | |
| AK060549.1 | - | 6 - 7 | 742 | 70 | ||
| AK100869.1 | - | 6 - 7 | 668 | 70 | ||
More Information
Set Membership: |
|

LOC4350083
Genomic Map