Assay Details
| Gene Symbol: LOC4350083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762471.2 | XP_015617957.1 | 8 - 9 | 1141 | 135 | |
| XM_015762472.2 | XP_015617958.1 | 8 - 9 | 1141 | 135 | ||
| GenBank mRNA | AK060326.1 | - | 8 - 9 | 1130 | 135 | |
| AK060549.1 | - | 8 - 9 | 1142 | 135 | ||
| AK100869.1 | - | 8 - 9 | 1068 | 135 | ||
More Information
Set Membership: |
|

LOC4350083
Genomic Map