Assay Details
| Gene Symbol: LOC4350086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761718.2 | XP_015617204.1 | 14 - 15 | 1804 | 75 | |
| GenBank mRNA | AK100578.1 | - | 12 - 13 | 2763 | 75 | |
More Information
Set Membership: |
|

LOC4350086
Genomic Map