Assay Details
| Gene Symbol: LOC4350086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761718.2 | XP_015617204.1 | 4 - 5 | 629 | 123 | |
| GenBank mRNA | AK100578.1 | - | 4 - 5 | 683 | 123 | |
More Information
Set Membership: |
|

LOC4350086
Genomic Map