Assay Details
| Gene Symbol: LOC4350619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760163.2 | XP_015615649.1 | 1 - 2 | 278 | 75 | |
| GenBank mRNA | AK059719.1 | - | 1 - 2 | 269 | 75 | |
More Information
Set Membership: |
|

LOC4350619
Genomic Map