Assay Details
| Gene Symbol: LOC4350865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761476.2 | XP_015616962.1 | 3 - 4 | 2919 | 87 | |
| GenBank mRNA | AK102379.1 | - | 3 - 4 | 2889 | 87 | |
More Information
Set Membership: |
|

LOC4350865
Genomic Map