Assay Details
| Gene Symbol: LOC4328406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770220.2 | XP_015625706.1 | 10 - 11 | 1838 | 96 | |
| GenBank mRNA | AK066725.1 | - | 10 - 11 | 1177 | 96 | |
| AK070569.1 | - | 10 - 11 | 1800 | 96 | ||
More Information
Set Membership: |
|

LOC4328406
Genomic Map