Assay Details
| Gene Symbol: LOC4328406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770220.2 | XP_015625706.1 | 1 - 2 | 865 | 155 | |
| GenBank mRNA | AK066725.1 | - | 1 - 2 | 205 | 155 | |
| AK070569.1 | - | 1 - 2 | 827 | 155 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770220.2 transcripts.
Set Membership: |
|

LOC4328406
Genomic Map