Assay Details
| Gene Symbol: LOC4328406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770220.2 | XP_015625706.1 | 9 - 10 | 1763 | 97 | |
| GenBank mRNA | AK066725.1 | - | 9 - 10 | 1102 | 97 | |
| AK070569.1 | - | 9 - 10 | 1725 | 97 | ||
More Information
Set Membership: |
|

LOC4328406
Genomic Map