Assay Details
| Gene Symbol: LOC4328422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770433.2 | XP_015625919.1 | 11 - 12 | 1675 | 83 | |
| XM_015770434.2 | XP_015625920.1 | 11 - 12 | 1600 | 83 | ||
| XM_015770435.2 | XP_015625921.1 | 11 - 12 | 1675 | 83 | ||
| GenBank mRNA | AK069161.1 | - | 10 - 11 | 1842 | 83 | |
More Information
Set Membership: |
|

LOC4328422
Genomic Map