Assay Details
| Gene Symbol: LOC4328457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771003.2 | XP_015626489.1 | 2 - 3 | 414 | 75 | |
| GenBank mRNA | AK060232.1 | - | 1 - 2 | 105 | 75 | |
More Information
Set Membership: |
|

LOC4328457
Genomic Map