Assay Details
| Gene Symbol: LOC4328501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767624.2 | XP_015623110.1 | 4 | 1435 | 89 | |
| GenBank mRNA | AK110722.1 | - | 1 | 951 | 89 | |
More Information
Set Membership: |
|

LOC4328501
Genomic Map