Assay Details
| Gene Symbol: LOC4328622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769944.2 | XP_015625430.1 | 1 - 2 | 342 | 105 | |
| GenBank mRNA | AK105298.1 | - | 1 - 2 | 327 | 105 | |
More Information
Set Membership: |
|

LOC4328622
Genomic Map