Assay Details
| Gene Symbol: LOC4328643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769181.2 | XP_015624667.1 | 4 - 5 | 531 | 148 | |
| XM_015769182.2 | XP_015624668.1 | 4 - 5 | 531 | 148 | ||
| GenBank mRNA | AK068100.1 | - | 4 - 5 | 452 | 148 | |
More Information
Set Membership: |
|

LOC4328643
Genomic Map