Assay Details
| Gene Symbol: LOC4328946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767149.2 | XP_015622635.1 | 1 - 2 | 4306 | 65 | |
| GenBank mRNA | AK073576.1 | - | 1 - 2 | 4313 | 65 | |
More Information
Set Membership: |
|

LOC4328946
Genomic Map