Assay Details
| Gene Symbol: LOC4328996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767351.1 | XP_015622837.1 | 4 - 5 | 505 | 59 | |
| GenBank mRNA | AK061802.1 | - | 1 - 2 | 844 | 59 | |
| AK099750.1 | - | 3 - 4 | 1941 | 59 | ||
More Information
Set Membership: |
|

LOC4328996
Genomic Map