Assay Details
| Gene Symbol: LOC4329266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770890.2 | XP_015626376.1 | 15 - 16 | 1658 | 70 | |
| GenBank mRNA | AK111848.1 | - | 13 - 14 | 1800 | 70 | |
More Information
Set Membership: |
|

LOC4329266
Genomic Map