Assay Details
| Gene Symbol: LOC4329266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770890.2 | XP_015626376.1 | 1 - 2 | 360 | 71 | |
| GenBank mRNA | AK111848.1 | - | 1 - 2 | 306 | 71 | |
More Information
Set Membership: |
|

LOC4329266
Genomic Map