Assay Details
| Gene Symbol: LOC4329266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770890.2 | XP_015626376.1 | 11 - 12 | 1265 | 83 | |
| GenBank mRNA | AK111848.1 | - | 9 - 10 | 1414 | 83 | |
More Information
Set Membership: |
|

LOC4329266
Genomic Map