Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 13 - 14 | 2164 | 69 | |
| GenBank mRNA | AK106285.1 | - | 9 - 10 | 1582 | 69 | |
| AK106581.1 | - | 1 - 2 | 142 | 69 | ||
| AY277897.2 | - | 13 - 14 | 1901 | 69 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map