Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 14 - 15 | 2281 | 71 | |
| GenBank mRNA | AK106285.1 | - | 10 - 11 | 1699 | 71 | |
| AY277897.2 | - | 14 - 15 | 2018 | 71 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map