Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 15 - 16 | 2415 | 79 | |
| GenBank mRNA | AK106285.1 | - | 11 - 12 | 1833 | 79 | |
| AK106581.1 | - | 3 - 4 | 391 | 79 | ||
| AY277897.2 | - | 15 - 16 | 2152 | 79 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map