Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 19 - 20 | 3207 | 66 | |
| GenBank mRNA | AK106285.1 | - | 15 - 16 | 2599 | 66 | |
| AK106581.1 | - | 7 - 8 | 1183 | 66 | ||
| AY277897.2 | - | 19 - 20 | 2944 | 66 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map