Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 20 - 21 | 3527 | 127 | |
| GenBank mRNA | AK106581.1 | - | 8 - 9 | 1503 | 127 | |
| AY277897.2 | - | 20 - 21 | 3264 | 127 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map