Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 21 - 22 | 3656 | 101 | |
| GenBank mRNA | AK106581.1 | - | 9 - 10 | 1632 | 101 | |
| AY277897.2 | - | 21 - 22 | 3393 | 101 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map