Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 25 - 26 | 4054 | 86 | |
| GenBank mRNA | AK106581.1 | - | 13 - 14 | 2030 | 86 | |
| AY277897.2 | - | 25 - 26 | 3791 | 86 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map