Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 6 - 7 | 1172 | 91 | |
| GenBank mRNA | AK106285.1 | - | 3 - 4 | 469 | 91 | |
| AY277897.2 | - | 6 - 7 | 909 | 91 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map