Assay Details
| Gene Symbol: LOC4329409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771294.2 | XP_015626780.1 | 7 - 8 | 1349 | 83 | |
| GenBank mRNA | AK106285.1 | - | 4 - 5 | 646 | 83 | |
| AY277897.2 | - | 7 - 8 | 1086 | 83 | ||
More Information
Set Membership: |
|

LOC4329409
Genomic Map