Assay Details
| Gene Symbol: LOC4329527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770882.2 | XP_015626368.1 | 4 - 5 | 1276 | 91 | |
| XM_015770883.2 | XP_015626369.1 | 4 - 5 | 1275 | 91 | ||
| GenBank mRNA | AK288723.1 | - | 4 - 5 | 1238 | 91 | |
More Information
Set Membership: |
|

LOC4329527
Genomic Map