Assay Details
| Gene Symbol: LOC4329892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769896.1 | XP_015625382.1 | 1 - 2 | 254 | 59 | |
| GenBank mRNA | AK062215.1 | - | 1 - 2 | 239 | 59 | |
| AK099086.1 | - | 1 - 2 | 223 | 59 | ||
| GQ848032.1 | - | 1 - 2 | 44 | 59 | ||
| KY038497.1 | - | 1 - 2 | 79 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769896.1 transcripts.
Set Membership: |
|

LOC4329892
Genomic Map