Assay Details
| Gene Symbol: LOC4329892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769896.1 | XP_015625382.1 | 2 - 3 | 275 | 114 | |
| GenBank mRNA | AK062215.1 | - | 2 - 3 | 260 | 114 | |
| AK099086.1 | - | 2 - 3 | 244 | 114 | ||
| GQ848032.1 | - | 2 - 3 | 65 | 114 | ||
| KY038497.1 | - | 2 - 3 | 100 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769896.1 transcripts.
Set Membership: |
|

LOC4329892
Genomic Map