Assay Details
| Gene Symbol: LOC4329892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769896.1 | XP_015625382.1 | 4 - 5 | 496 | 109 | |
| GenBank mRNA | AK099086.1 | - | 4 - 5 | 465 | 109 | |
| GQ848032.1 | - | 4 - 5 | 286 | 109 | ||
| KY038497.1 | - | 4 - 5 | 321 | 109 | ||
More Information
Set Membership: |
|

LOC4329892
Genomic Map