Assay Details
| Gene Symbol: LOC4329892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769896.1 | XP_015625382.1 | 9 - 10 | 1046 | 87 | |
| GenBank mRNA | AF546879.1 | - | 5 - 6 | 488 | 87 | |
| AK062215.1 | - | 9 - 10 | 1030 | 87 | ||
| AK099086.1 | - | 9 - 10 | 1015 | 87 | ||
| BT131063.1 | - | 4 - 5 | 454 | 87 | ||
| GQ848032.1 | - | 9 - 10 | 836 | 87 | ||
| KY038497.1 | - | 9 - 10 | 871 | 87 | ||
More Information
Set Membership: |
|

LOC4329892
Genomic Map