Assay Details
| Gene Symbol: LOC4329905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767821.2 | XP_015623307.1 | 1 - 2 | 208 | 101 | |
| GenBank mRNA | AK068016.1 | - | 1 - 2 | 172 | 101 | |
| AK068109.1 | - | 1 - 2 | 189 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767821.2 transcripts.
Set Membership: |
|

LOC4329905
Genomic Map