Assay Details
| Gene Symbol: LOC4330248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769907.1 | XP_015625393.1 | 2 - 3 | 762 | 79 | |
| GenBank mRNA | AB009665.1 | - | 2 - 3 | 689 | 79 | |
| AK061769.1 | - | 2 - 3 | 691 | 79 | ||
| AK104658.1 | - | 2 - 3 | 691 | 79 | ||
| EU267998.1 | - | 2 - 3 | 670 | 79 | ||
| JN208223.1 | - | 2 - 3 | 663 | 79 | ||
| JQ823126.1 | - | 2 - 3 | 634 | 79 | ||
| JQ823127.1 | - | 2 - 3 | 634 | 79 | ||
| KF737329.1 | - | 2 - 3 | 670 | 79 | ||
| KT862204.1 | - | 2 - 3 | 663 | 79 | ||
More Information
Set Membership: |
|

LOC4330248
Genomic Map