Assay Details
| Gene Symbol: LOC4330297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770130.2 | XP_015625616.1 | 1 - 2 | 545 | 129 | |
| GenBank mRNA | AB101648.1 | - | 1 - 2 | 231 | 129 | |
| AK111544.1 | - | 1 - 2 | 551 | 129 | ||
More Information
Set Membership: |
|

LOC4330297
Genomic Map