Assay Details
| Gene Symbol: LOC4330297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770130.2 | XP_015625616.1 | 7 - 8 | 2135 | 81 | |
| GenBank mRNA | AB101648.1 | - | 7 - 8 | 1821 | 81 | |
| AK111544.1 | - | 7 - 8 | 2141 | 81 | ||
More Information
Set Membership: |
|

LOC4330297
Genomic Map