Assay Details
| Gene Symbol: LOC4330322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772164.2 | XP_015627650.1 | 1 - 2 | 264 | 76 | |
| XM_015772165.2 | XP_015627651.1 | 1 - 2 | 261 | 76 | ||
| GenBank mRNA | AK103328.1 | - | 1 - 2 | 237 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772164.2, XM_015772165.2 transcripts.
Set Membership: |
|

LOC4330322
Genomic Map