Assay Details
| Gene Symbol: LOC4330322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772164.2 | XP_015627650.1 | 2 - 3 | 1111 | 109 | |
| XM_015772165.2 | XP_015627651.1 | 2 - 3 | 1108 | 109 | ||
| GenBank mRNA | AK103328.1 | - | 2 - 3 | 1084 | 109 | |
More Information
Set Membership: |
|

LOC4330322
Genomic Map