Assay Details
| Gene Symbol: LOC4330322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772164.2 | XP_015627650.1 | 6 - 7 | 1805 | 87 | |
| XM_015772165.2 | XP_015627651.1 | 5 - 6 | 1694 | 87 | ||
| GenBank mRNA | AK103328.1 | - | 6 - 7 | 1778 | 87 | |
More Information
Set Membership: |
|

LOC4330322
Genomic Map