Assay Details
| Gene Symbol: LOC4330380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772113.2 | XP_015627599.1 | 7 - 8 | 1171 | 99 | |
| GenBank mRNA | AK101568.1 | - | 7 - 8 | 1144 | 99 | |
| AK105916.1 | - | 7 - 8 | 1167 | 99 | ||
More Information
Set Membership: |
|

LOC4330380
Genomic Map