Assay Details
| Gene Symbol: LOC4330388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770605.2 | XP_015626091.1 | 3 - 4 | 578 | 124 | |
| GenBank mRNA | AK069300.1 | - | 3 - 4 | 460 | 124 | |
More Information
Set Membership: |
|

LOC4330388
Genomic Map