Assay Details
| Gene Symbol: LOC4330431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768271.1 | XP_015623757.1 | 1 - 2 | 595 | 70 | |
| GenBank mRNA | AK059448.1 | - | 1 - 2 | 600 | 70 | |
More Information
Set Membership: |
|

LOC4330431
Genomic Map