Assay Details
| Gene Symbol: LOC4330431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768271.1 | XP_015623757.1 | 5 - 6 | 872 | 85 | |
| GenBank mRNA | AK059448.1 | - | 5 - 6 | 877 | 85 | |
More Information
Set Membership: |
|

LOC4330431
Genomic Map