Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 12 - 13 | 2907 | 98 | |
| XM_015769940.2 | XP_015625426.1 | 11 - 12 | 2670 | 98 | ||
| GenBank mRNA | AB477099.1 | - | 11 - 12 | 2593 | 98 | |
| AK121261.1 | - | 12 - 13 | 2907 | 98 | ||
| AK121875.1 | - | 12 - 13 | 2862 | 98 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map