Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 17 - 18 | 3500 | 74 | |
| XM_015769940.2 | XP_015625426.1 | 16 - 17 | 3263 | 74 | ||
| GenBank mRNA | AB477099.1 | - | 16 - 17 | 3186 | 74 | |
| AK121261.1 | - | 17 - 18 | 3500 | 74 | ||
| AK121875.1 | - | 17 - 18 | 3455 | 74 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map